Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lab Vortex Mixers with Touch Function

Lab Vortex Mixers with Touch Function

110 - 120 V, 50/60 Hz, 96 W (US/NA), 0.02 ml, 0.5 ml, 1.5/2.0 ml, 5 ml, 15 ml, 50 ml and Standard assay plate
Catalog # 76027-828
Irregular Voltage
Supplier:   Heathrow Scientific
Supplier #:  HS120209
Lab Vortex Mixers with Touch Function
Catalog # 76027-828
Irregular Voltage
Lab Vortex Mixers with Touch Function
Lab Vortex Mixers with Touch Function

110 - 120 V, 50/60 Hz, 96 W (US/NA), 0.02 ml, 0.5 ml, 1.5/2.0 ml, 5 ml, 15 ml, 50 ml and Standard assay plate

Price
$626.21
Log in for availability
Each
Product Details & Documents

This unique and innovative designed Vortex Mixer is a variable speed vortexing unit with multi-purpose and maxium convenience. Why take time to search for separate mixing heads for various sized tubes when this vortex mixer provides one mixing head that will accomodate tubes ranging from 0.2 mL - 50 mL and a standard assay plate.

  • Variable speed rotary dial provides pre-set speed control (1000, 2000, 3000 rpm) during vortex mixing
  • Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
  • Innovative head can be removed for cleaning purposes
  • High impact ABS body shell
  • US patent

This sleek and modern designed vortex mixer can meet the mixing requirements of the most popular test tubes used in the today’s lab without the need to change mixing head units.

The innovative design makes maximum use of space available providing holding slots for no less than six different sizes of tubes and a standard assay plate, while the center pad area can be used for quick hand-held mixing of a vast range of tubes and vessels.

Certifications: SGS-US-C, CE, CB Scheme, RoHS 2, PSE, and WEEE compliant.

Ordering information: Five-year warranty.

Delivery information: The Vortexer comes with a mixing head attachment, three central intermittent vortexing pads, three elastic retention cords, and a power cord.

Packaging: Plain white box with label.

Documents

Specifications
  • Capacity:
    0.02 ml, 0.5 ml, 1.5/2.0 ml, 5 ml, 15 ml, 50 ml and Standard assay plate
  • Electrical:
    110 - 120 V, 50/60 Hz, 96 W (US/NA)
  • Cat. No.:
    76027-828
  • Speed range:
    1000 - 3000 rpm
  • Weight:
    8.6 lbs.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.