Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REAGENT POTASSIUM PERMANGANATE ACS 25 G
103479-552
:
Aqua Solutions
REAGENT POTASSIUM PERMANGANATE ACS 25 G
103479-552
:
Aqua Solutions
:
P5450-25G
ghs_P5450