Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Solution 2 Lysis Buffer, Zymo Research

Solution 2 Lysis Buffer, Zymo Research

90 mL
Catalog # 76020-308
Supplier:   Zymo Research
Supplier #:  D2004-2-90
Solution 2 Lysis Buffer, Zymo Research
Catalog # 76020-308
Supplier:  Zymo Research
CAS Number:  
Solution 2 Lysis Buffer, Zymo Research
Solution 2 Lysis Buffer, Zymo Research

90 mL

CAS Number:  
Price
$227.78
Log in for availability
Each (90ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Size:
    90 mL
  • Brand Name:
    Zymo Research
  • Cat. No.:
    76020-308

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.