Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

ProPette™ Electronic Pipette Controller

Filters (0.45 µm)
Catalog # 470228-484
Supplier:   MTC BIO, INC.
 :  P6080-FI
ProPette™ Electronic Pipette Controller
Catalog # 470228-484
Supplier:  MTC BIO, INC.
ProPette™ Electronic Pipette Controller
ProPette™ Electronic Pipette Controller

Filters (0.45 µm)

Price
$55.90
Log in for availability
Each Pack of 5
Product Details & Documents

ProPette™ features an ergonomic body design, with computer-balanced weight distribution and low pressure fingertip control buttons that combine to provide a more comfortable pipetting experience.

  • QuickStand™ prevents pipette contamination
  • Variable speed control
  • Cordless and rechargable
  • Provides 8 hours of continuous operation
  • Fits serological pipettes from 1 to 100 ml
  • Ambidexterous

Retractable Quickstand™ allows the instrument to be set down on the bench without the risk of contaminating pipettes.

Specifications
  • Description:
    Filters (0.45 µm)