Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Phosphate Buffer

Phosphate Buffer

Phosphate buffer, 1 L
Catalog # 76019-252
Supplier:   Hardy Diagnostics
Supplier #:  U192
Phosphate Buffer
Catalog # 76019-252
Supplier:  Hardy Diagnostics
CAS Number:  
Phosphate Buffer
Phosphate Buffer

Phosphate buffer, 1 L

CAS Number:  
Price
$484.00
Log in for availability
Pack of 10 (1l)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Documents

Specifications

Specifications

  • Description:
    Phosphate buffer
  • Volume:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.