Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hardshell Plastic Cuvette Cases, Type P

Hardshell Plastic Cuvette Cases, Type P

Cuvette case for 45 mm cells, Polyfoam
Catalog # 76016-134
Hardshell Plastic Cuvette Cases, Type P
Catalog # 76016-134
Supplier:  
Hardshell Plastic Cuvette Cases, Type P
Hardshell Plastic Cuvette Cases, Type P

Cuvette case for 45 mm cells, Polyfoam

Discontinued
Product Details & Documents

Cuvette cases hold 4 cells up to 10 mm L/P each, or two 20 mm L/P cells, or one 50 mm L/P cell.

  • Black opaque finish
Specifications
  • Description:
    Cuvette case for 45 mm cells
  • Material:
    Polyfoam
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.