Orders LinkContactUsLinkComponent
Cuvette Washer, FireflySci
Single Cuvette Washer Kit
  76015-788
 :  P65SKIT
Cuvette Washer, FireflySci
  76015-788
Cuvette Washer, FireflySci
Cuvette Washer, FireflySci
Single Cuvette Washer Kit
$704.79
Each
Cuvette washers are handmade and built to last for multiple uses without contamination.
  • Fast cuvette cleaning action
  • Leave cuvettes in place for quick drying action
  • Use with cuvettes from 3 to10mm lightpath
  • Reduces handling and risk of breakage
Easy to use device requires the cuvette to be turned upside down when the cleaning solution is run.
 : Washers includes Gasket and stopper. Vacuum flask and pump must be ordered separately. Double cuvette washer kit includes cuvette washer head, rubber gasket, stopper (size 7), filtering flask, and vinyl tubing (304.8 cm). Kits are also available with vacuum pump.
  • Description:
    Single Cuvette Washer Kit
Accessories
  •  
    Description
  • 76015-792
    Small Rubber Gasket for Single & Double Cuvette Washer
    Each - $24.15
  •  
    Description
  • 76015-794
    Size 7 Vacuum Flask Stopper for Cuvette Washers
    Each - $53.01