Orders LinkContactUsLinkComponent
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
UV Quartz, 100 µl (16×), 10 mm
Catalog # 76015-216
Supplier:   FIREFLYSCI, INC.
Supplier #:  16UV10
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Catalog # 76015-216
Supplier:  FIREFLYSCI, INC.
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
UV Quartz, 100 µl (16×), 10 mm
Price
$2,485.34
Log in for availability
Each
Product Details & Documents
Multi-Micro Cell Array has 16-chambered multi-micro array of cells for doing multiple measurements.
  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Fire-fused to ensure that cuvettes are robust and durable
  • Comes in UV Quartz
  • Available in 10 mm lightpath
  • Two windows optically polished
  • Walls are made from black quartz to block out stray light
Typically used in Shimadzu machines.
Each cell goes through a rigorous inspection process before leaving the factory.
Specifications
  • Volume:
    100 µl (16×)
  • Description:
    UV Quartz
  • Lightpath:
    10 mm