Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade, Spectrum Chemicals

Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade, Spectrum Chemicals

Phosphate Buffered Saline (PBS) Solution, 1X, Sterile, BiotechGrade, 500 ml, Poly bottle, 7.3 - 7.5
Catalog # 76014-826
Supplier #:  P3033-500ML
Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade, Spectrum Chemicals
Catalog # 76014-826
CAS Number:  
Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade, Spectrum Chemicals

Phosphate Buffered Saline (PBS) Solution, 1X, Sterile, BiotechGrade, 500 ml, Poly bottle, 7.3 - 7.5

CAS Number:  
Price
$204.91
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade is abbreviated as PBS and is used in biological research as a buffer solution containing sodium chloride and sodium phosphate, as well as potassium phosphate and potassium chloride in some formulations.

  • Sterile
  • BiotechGrade
  • DNase: none detected

It is also used in cell container rinsing and substance dilution.

Documents

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Description:
    Phosphate Buffered Saline (PBS) Solution, 1X, Sterile, BiotechGrade
  • pH:
    7.3 - 7.5
  • Volume:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.