Storage Bins
Shelf bin set (Six SOVE3010-6 bins), Blue, PP, Pack of 6, 11.81×3.62×3.23"
Catalog # 76015-064
Supplier: Treston
Supplier #: SBS6-3010-6
Jump to:
Product Details & Documents
These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.
- Straight sides ensure an efficient use of space
- Multiple color options make finding the right item easy
- Open front provides easy access
- Corrugated base helps retrieve contents, especially smaller sized and flat objects
Ordering information: Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.
Specifications
- Color:Blue
- Description:Shelf bin set (Six SOVE3010-6 bins)
- Dimensions:11.81×3.62×3.23"
- Material:PP
- Size:Pack of 6



