Storage Bins
Stacking bin set (Four SOVE1015-6 bins), Blue, PP, Pack of 4, 6.5×4.13×2.95"
Catalog # 76015-060
Supplier: Treston
Supplier #: SBS4-1015-6
Jump to:
Product Details & Documents
These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.
- Straight sides ensure an efficient use of space
- Multiple color options make finding the right item easy
- Open front provides easy access
- Corrugated base helps retrieve contents, especially smaller sized and flat objects
Ordering information: Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.
Specifications
- Color:Blue
- Description:Stacking bin set (Four SOVE1015-6 bins)
- Dimensions:6.5×4.13×2.95"
- Material:PP
- Size:Pack of 4



