Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Storage Bins

ESD stacking bin set (Two SOVE1525-6 and two SOVE1520-6), Black, PP, Pack of 4, (2) 9.84×5.87×5.12" and (2) 7.56×5.87×4.13"
  76015-034
 :   Treston
 :  SBS4-2ESD
Storage Bins
  76015-034
 :  Treston
Storage Bins
Storage Bins

ESD stacking bin set (Two SOVE1525-6 and two SOVE1520-6), Black, PP, Pack of 4, (2) 9.84×5.87×5.12" and (2) 7.56×5.87×4.13"

 
$90.91
   
Pack of 1
 
 

These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.

  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects

 : Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.

 
  • Color:
    Black
  • Description:
    ESD stacking bin set (Two SOVE1525-6 and two SOVE1520-6)
  • Dimensions:
    (2) 9.84×5.87×5.12" and (2) 7.56×5.87×4.13"
  • Material:
    PP
  • Size:
    Pack of 4