Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Storage Bins

ESD stacking bin set (Four SOVE1015-4ESD bins), Black, PP, Pack of 4, 6.5×4.13×2.95"
Catalog # 76015-032
Supplier:   Treston
 :  SBS4-1015-4ESD
Storage Bins
Catalog # 76015-032
Supplier:  Treston
Storage Bins
Storage Bins

ESD stacking bin set (Four SOVE1015-4ESD bins), Black, PP, Pack of 4, 6.5×4.13×2.95"

Price
$34.12
Log in for availability
Pack of 1
 
Product Details & Documents

These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.

  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects

 : Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.

Specifications
  • Color:
    Black
  • Description:
    ESD stacking bin set (Four SOVE1015-4ESD bins)
  • Dimensions:
    6.5×4.13×2.95"
  • Material:
    PP
  • Size:
    Pack of 4