Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ASSAY MOUSE IGM/IGG DC 96W WHITE 2 PLATE
Catalog # MSPP-MIGMIGGDC
Supplier:
CELLULAR TECHNOLOGY LIMITED MS
Expand image
ASSAY MOUSE IGM/IGG DC 96W WHITE 2 PLATE
Catalog # MSPP-MIGMIGGDC
Supplier:
CELLULAR TECHNOLOGY LIMITED MS
Supplier Number:
MIGMIGG-DCE-1M2