$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
ASSAY MOUSE IGA SC 96WELL WHITE 10 PLATE
MSPP-MIGASCE1M
:
CELLULAR TECHNOLOGY LIMITED MS
ASSAY MOUSE IGA SC 96WELL WHITE 10 PLATE
MSPP-MIGASCE1M
:
CELLULAR TECHNOLOGY LIMITED MS
:
MIGA-SCE-1M10