Various Replacement Parts, Sciencix
Capillary, Loop, 100 µl, 1100, 1120, 1220, G1313A, G1329A/B, AGLS, 01078-87302
Catalog # 103383-936
Marketsource
Supplier: SCIENCIX
: 11-4059
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Capillary, Loop, 100 µl
- OEM manufacturer:AGLS
- OEM equivalent:01078-87302
- Model:1100, 1120, 1220, G1313A, G1329A/B