CRYOPRESERVATION MEDIUM CS5
Catalog # MSPP-07933
Supplier: Stemcell Technologies
CRYOPRESERVATION MEDIUM CS5
Catalog # MSPP-07933
Supplier: Stemcell Technologies
Supplier Number: 07933
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Disposable Serological Pipettes
Supplier: VWR International
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Corning® 96-well Clear Polystyrene Microplates, Corning
Supplier: Corning
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
REAGENT HPSC ENZYMEFREE PASSAGING 100ML
Supplier: Stemcell Technologies