Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Titanium Micro Forceps, Sklar

Titanium Micro Forceps, Sklar

Titanium, 178 mm (7"), 1 x 0.5 mm
Catalog # 75995-940
Supplier:   Sklar
Supplier #:  98-4331T
Titanium Micro Forceps, Sklar
Catalog # 75995-940
Supplier:  Sklar
Titanium Micro Forceps, Sklar
Titanium Micro Forceps, Sklar

Titanium, 178 mm (7"), 1 x 0.5 mm

Price
$3,193.04
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

All Sklar instruments are industry standard.

  • Reusable
  • Nonsterile
Specifications
  • Length:
    178 mm (7")
  • Material:
    Titanium
  • Tip dimensions:
    1 x 0.5 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.