Titanium Micro Forceps, Sklar
Product Details & Documents
All Sklar instruments are industry standard.
- Reusable
- Nonsterile
Specifications
- Length:152 mm (6")
- Material:Titanium
- Tip dimensions:1 x 0.5 mm