Objectives for REVOLVE Microscopes, Echo
Achromat objective, 18.5 mm, 0.10 NA
Catalog # 75992-492
Jump to:
Discontinued
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:4×
- Description:Achromat objective
- Aperture:0.10 NA
- Working distance:18.5 mm