Objectives for REVOLVE Microscopes, Echo
Apochromat objective, 6.2 mm, 0.08 NA
Catalog # 75992-482
Supplier: DISCOVER ECHO INC.
: JMAG0020
Jump to:
Product Details & Documents
Specifications
- Magnification:2×
- Description:Apochromat objective
- Aperture:0.08 NA
- Working distance:6.2 mm