Objectives for REVOLVE Microscopes, Echo
Achromat phase ELWD objective, 2.2 mm, 0.55 NA
Catalog # 75990-672
Supplier: DISCOVER ECHO INC.
: JMAG0409
Jump to:
Product Details & Documents
Specifications
- Magnification:40×
- Description:Achromat phase ELWD objective
- Aperture:0.55 NA
- Working distance:2.2 mm