Objectives for REVOLVE Microscopes, Echo
Fluorite objective, 0.51 mm, 0.75 NA
Catalog # 75990-664
Supplier #: JMAG0405
Jump to:
Discontinued
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:40×
- Description:Fluorite objective
- Aperture:0.75 NA
- Working distance:0.51 mm