Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD objective, 2.7 - 4 mm, 0.60 NA
Catalog # 75990-660
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0403
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:40×
- Description:Fluorite ELWD objective
- Aperture:0.60 NA
- Working distance:2.7 - 4 mm