CELL NRK RAT KIDNEY EPITHELIAL OSB-MEND
Catalog # MSPP-CRL6509
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
CELL NRK RAT KIDNEY EPITHELIAL OSB-MEND
Catalog # MSPP-CRL6509
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: CRL-6509
Frequently Bought Together
Static Mouse Cage, 1284L
Supplier: Tecniplast Usa
Reusable Plastic Animal Cages, Tecniplast®
Supplier: Tecniplast Usa
Ward's® Bulk Simulated Antisera
Supplier: Avantor
Tricine, Ultrapure
Supplier: VWR
VWR® Shelf Liners
Supplier: VWR International
neodisher® Detergents, Miele
Supplier: Miele
Bleomycin sulfate (mixture) ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Supplier: Thermo Fisher Scientific
PVC Film Wrap, T FRANK MCCALLS INC
Supplier: TFrank McCalls
KIT CUSTOM CUT 2-LAYER DRAWER LINER FOAM
Supplier: GLOBAL EQUIPMENT MS
COVER SHOE STANDARD SZ 12-15 CS150
Supplier: ULINE MS
SUTURE V20 2-0 15CM CS12
Supplier: CIA MEDICAL MS
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Biodyne™ B Nylon Membrane, 0.45 µm, Thermo Scientific
Supplier: Invitrogen
CELL LINE RF/6A (ATCC CRL-1780)
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Static Rat/Hamster Cages, 2154F
Supplier: Tecniplast Usa