Nitrate Substrate Broth, Hardy Diagnostics
AFB Nitrate Reagent B, 15 ml, —
Supplier: Hardy Diagnostics
Supplier #: Z125
Nitrate Substrate Broth, Hardy Diagnostics
Catalog # 75993-560
Supplier: Hardy Diagnostics
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
For the determination of nitrate reactions of mycobacteria. Use with AFB Nitrate Reagent A and B
Reagent A contains sulfanilimide, 0.2%.
Reagent A contains sulfanilimide, 0.2%
Reagent B contains naphthylethylenediamine 2HCL, 0.1%
Reagent B contains naphthylethylenediamine 2HCL, 0.1%.
Documents
Specifications
Specifications
- Description:AFB Nitrate Reagent B
- Minimum Order Quantity:—
- Fill volume:15 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



