Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Nitrate Substrate Broth, Hardy Diagnostics

Nitrate Substrate Broth, Hardy Diagnostics

Nitrate Substrate Broth, 13×100 mm, 2 ml, 5 packs, 13 x 100 mm, PC tube, optically clear, shatter resistant
Catalog # 75993-554
Supplier:   Hardy Diagnostics
Supplier #:  R97
Nitrate Substrate Broth, Hardy Diagnostics
Catalog # 75993-554
Supplier:  Hardy Diagnostics
CAS Number:  
Nitrate Substrate Broth, Hardy Diagnostics
Nitrate Substrate Broth, Hardy Diagnostics

Nitrate Substrate Broth, 13×100 mm, 2 ml, 5 packs, 13 x 100 mm, PC tube, optically clear, shatter resistant

CAS Number:  
Price
$131.00
Log in for availability
Pack of 20 (2ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For the determination of nitrate reactions of mycobacteria. Use with AFB Nitrate Reagent A and B

Reagent A contains sulfanilimide, 0.2%.

Reagent A contains sulfanilimide, 0.2%
Reagent B contains naphthylethylenediamine 2HCL, 0.1%

Reagent B contains naphthylethylenediamine 2HCL, 0.1%.

Specifications

Specifications

  • Pack type:
    PC tube, optically clear, shatter resistant
  • Description:
    Nitrate Substrate Broth, 13×100 mm
  • Size:
    13 x 100 mm
  • Minimum Order Quantity:
    5 packs
  • Fill volume:
    2 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.