Objectives for REVOLVE Microscopes, Echo
Achromat SLWD objective, 25 mm, 0.25 NA
Catalog # 75990-652
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0208
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:20×
- Description:Achromat SLWD objective
- Aperture:0.25 NA
- Working distance:25 mm