Orders LinkContactUsLinkComponent
Objectives for REVOLVE Microscopes, Echo
Objectives for REVOLVE Microscopes, Echo
Fluorite phase objective, 2.1 mm, 0.50 NA
Catalog # 75990-646
Supplier #:  JMAG0205
Objectives for REVOLVE Microscopes, Echo
Catalog # 75990-646
Supplier:  
Objectives for REVOLVE Microscopes, Echo
Fluorite phase objective, 2.1 mm, 0.50 NA
Discontinued
Product Details & Documents
Documents
Specifications
  • Magnification:
    20×
  • Description:
    Fluorite phase objective
  • Aperture:
    0.50 NA
  • Working distance:
    2.1 mm