Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD phase objective, 6.6 - 7.8 mm, 0.45 NA
Catalog # 75990-642
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0203
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:20×
- Description:Fluorite ELWD phase objective
- Aperture:0.45 NA
- Working distance:6.6 - 7.8 mm