Orders LinkContactUsLinkComponent
Objectives for REVOLVE Microscopes, Echo
Objectives for REVOLVE Microscopes, Echo
Plan fluorite LWD CC, 6.6 - 7.8 mm, 0.45 NA
Catalog # 75990-640
Supplier:   DISCOVER ECHO INC.
Supplier #:  JMAG0202
Objectives for REVOLVE Microscopes, Echo
Catalog # 75990-640
Supplier:  DISCOVER ECHO INC.
Objectives for REVOLVE Microscopes, Echo
Plan fluorite LWD CC, 6.6 - 7.8 mm, 0.45 NA
Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Documents
Specifications
  • Magnification:
    20×
  • Description:
    Plan fluorite LWD CC
  • Aperture:
    0.45 NA
  • Working distance:
    6.6 - 7.8 mm