Orders LinkContactUsLinkComponent
Objectives for REVOLVE Microscopes, Echo
Achromat ELWD phase objective, 9.8 mm, 0.25 NA
Catalog # 75990-636
 :  JMAG0106
Objectives for REVOLVE Microscopes, Echo
Catalog # 75990-636
Supplier:  
Objectives for REVOLVE Microscopes, Echo
Achromat ELWD phase objective, 9.8 mm, 0.25 NA
Discontinued
Product Details & Documents
Documents
Specifications
  • Magnification:
    10×
  • Description:
    Achromat ELWD phase objective
  • Aperture:
    0.25 NA
  • Working distance:
    9.8 mm