Orders LinkContactUsLinkComponent
Objectives for REVOLVE Microscopes, Echo
Objectives for REVOLVE Microscopes, Echo
Plan fluorite, 10 mm, 0.30 NA
Catalog # 75990-628
Supplier #:  JMAG0102
Objectives for REVOLVE Microscopes, Echo
Catalog # 75990-628
Supplier:  
Objectives for REVOLVE Microscopes, Echo
Plan fluorite, 10 mm, 0.30 NA
Discontinued
Product Details & Documents
Documents
Specifications
  • Magnification:
    10×
  • Description:
    Plan fluorite
  • Aperture:
    0.30 NA
  • Working distance:
    10 mm