Objectives for REVOLVE Microscopes, Echo
Fluorite oil phase objective, 0.2 mm, 1.30 NA
Catalog # 75990-530
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG1004
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:100×
- Description:Fluorite oil phase objective
- Aperture:1.30 NA
- Working distance:0.2 mm