Objectives for REVOLVE Microscopes, Echo
Achromat objective, 0.17 mm, 0.80 NA
Catalog # 75990-522
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0608
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:60×
- Description:Achromat objective
- Aperture:0.80 NA
- Working distance:0.17 mm