Objectives for REVOLVE Microscopes, Echo
Fluorite oil objective, 0.12 mm, 1.25 - 0.65 NA
Catalog # 75990-518
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0606
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:60×
- Description:Fluorite oil objective
- Aperture:1.25 - 0.65 NA
- Working distance:0.12 mm