Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD Phase objective, 1.5 - 2.2 mm, 0.70 NA
Catalog # 75990-516
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0605
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:60×
- Description:Fluorite ELWD Phase objective
- Aperture:0.70 NA
- Working distance:1.5 - 2.2 mm