$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
ASSAY MOUSE CREATININE ENZYMATIC 96TEST
MSPP-80350
:
CRYSTAL CHEM INC MS
ASSAY MOUSE CREATININE ENZYMATIC 96TEST
MSPP-80350
:
CRYSTAL CHEM INC MS
:
80350