Orders LinkContactUsLinkComponent
PARK7
PARK7
50 µg
Catalog # 75930-734
PARK7
Catalog # 75930-734
Supplier:  
CAS Number:  
PARK7
50 µg
CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
PARK7(DJ-1) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PARK7 or parkinson protein 7 belongs to the peptidase C56 family of proteins which acts as a positive regulator of androgen receptor-dependent transcription. PARK7 also functions as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. PARK7 mutations that impair transcriptional co-activator function can render dopaminergic neurons vulnerable to apoptosis and may contribute to the pathogenesis of Parkinson disease (1). PARK7 is an atypical peroxiredoxin-like peroxidase that scavenges hydrogen peroxide through oxidation of cys106 (2). PARK7 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, and Neurobiology research.
PARK7 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. PARK7 Protein is suitable for use in Western Blot. Expect a band approximately ~22kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Specifications
Specifications
  • Size:
    50 µg
  • Cat. No.:
    75930-734
  • Shipping Temperature:
    Dry Ice
  • Protein Synonyms:
    PARK7|DJ-1
  • Protein/Peptide Name:
    PARK7