EpiScript™ RNase H- Reverse Transcriptase, cDNA synthesis, Biosearch Technologies
EpiScript™ RNase H- Reverse Transcriptase, 10000 Units
75927-940
: Lucigen
Supplier #: ERT12910K
EpiScript™ RNase H- Reverse Transcriptase, cDNA synthesis, Biosearch Technologies
75927-940
: Lucigen
:
An optimized MMLV reverse transcriptase enzyme for improved production of full-length cDNA from limiting total RNA samples.
- Efficient and Sensitive: Produce full-length cDNA from as little as 50 pg of total RNA
- Improved Specificity: Run reaction temperatures as high as 55°C to enhance primer specificity and improve transcription of GC-rich or high secondary structure target RNAs
EpiScript™ RNase H- Reverse Transcriptase (EpiScript RT), an alternative to SuperScript® II Reverse Transcriptase, is a recombinant MMLV reverse transcriptase with greatly reduced RNase H activity. It is highly efficient at producing full-length cDNA from long RNA templates and is capable of producing cDNA from as little as 50 pg of total RNA for real-time RT-PCR (qRT-PCR) analysis and other applications.
Delivery information: Contains EpiScript RNase H- Reverse Transcriptase, 10X Reaction Buffer and 100 mM DTT.
Caution: For research use only. Not for human or diagnostic use.
Documents
MA353E EpiScript-RT
Manual
- Description:EpiScript™ RNase H- Reverse Transcriptase
- Size:10000 Units