Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REAGENT BCP PHASE SEPARATION 200ML
Catalog # MSPP-BP151
Supplier:
MOLECULAR RESEARCH CENTER I MS
SDS
Expand image
REAGENT BCP PHASE SEPARATION 200ML
Catalog # MSPP-BP151
Supplier:
MOLECULAR RESEARCH CENTER I MS
Supplier Number:
BP-151
SDS
Related Information
Documentation
SDS
ghs_BP-151.pdf
Documentation