Iodine ≥99.8%, solid ACS, Fluka™
Glass bottle, 2.5 kg
Supplier: Honeywell Research Chemicals
Supplier #: 207772-2.5KG
Iodine ≥99.8%, solid ACS, Fluka™
Catalog # BJ207772-2.5KG
Supplier: Honeywell Research Chemicals
CAS Number: 7553-56-2
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Honeywell ACS solvents for general laboratory use are:
- A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested tmeet ACS general use specifications
- CAS Number:7553-56-2
- Formula:I₂
- MDL Number:MFCD00011355
- Molecular Weight:253.81 g/mol
- Melting Pt:113.5 °C
- Density:4.930 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-442-4
- UN:3495
- ADR:8,III
Documents
Specifications
Specification Test Results
- Assay:Min. 99.8 %
- Non-volatile matter:Max. 0.01 %
- Bromine and chlorine (as chloride):Max. 0.005 %
- Form:Solid
- Vapor Pressure:0.31 mmHg ( 25 °C)
- Resistivity:1.3E15 μΩ-cm
Specifications
- Pack type:Glass bottle
- Size:2.5 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



