Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Iodine ≥99.8%, solid ACS, Fluka™

Iodine ≥99.8%, solid ACS, Fluka™

Glass bottle, 1 kg
Catalog # BJ207772-1KG
Health
Irritant
Supplier #:  207772-1KG
Iodine ≥99.8%, solid ACS, Fluka™
Catalog # BJ207772-1KG
CAS Number:  7553-56-2
Iodine ≥99.8%, solid ACS, Fluka™

Glass bottle, 1 kg

CAS Number:  7553-56-2
Price
$761.15
Log in for availability
Each (1kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Honeywell ACS solvents for general laboratory use are:

  • A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested tmeet ACS general use specifications
  • CAS Number:7553-56-2
  • Formula:I₂
  • MDL Number:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Melting Pt:113.5 °C
  • Density:4.930 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-442-4
  • UN:3495
  • ADR:8,III
Specifications

Specification Test Results

  • Assay:
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.01 %
  • Bromine and chlorine (as chloride):
    Max. 0.005 %
  • Form:
    Solid
  • Vapor Pressure:
    0.31 mmHg ( 25 °C)
  • Resistivity:
    1.3E15 μΩ-cm

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.