Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
(±)-2-Butanol

(±)-2-Butanol

Glass, amber, 1 L
Catalog # 75871-922
Supplier #:  B1187-1LTGL
(±)-2-Butanol
Catalog # 75871-922
CAS Number:  78-92-2
(±)-2-Butanol
(±)-2-Butanol

Glass, amber, 1 L

CAS Number:  78-92-2
Price
$169.29
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

sec-Butyl Alcohol is also known as 2-Butyl alcohol and sec-Butanol. It is used as a solvent and can be converted to butanone which is an industrial solvent.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
  • CAS Number:78-92-2
  • Formula:H₃CCH₂CH(OH)CH₃
  • MDL Number:MFCD00004569
  • Molecular Weight:74.12 g/mol
  • Density:0.808
  • Storage Temperature:Ambient
  • UN:1120
  • ADR:3,III

Documents

Specifications

Specifications

  • Pack type:
    Glass, amber
  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.