Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Hitachi M4 SEM Mount, Electron Microscopy Sciences

SEM Mount Hitachi M4 Aluminium 25 mm, 50/pack
  103306-258
 :  75662-20
Hitachi M4 SEM Mount, Electron Microscopy Sciences
  103306-258
Hitachi M4 SEM Mount, Electron Microscopy Sciences

SEM Mount Hitachi M4 Aluminium 25 mm, 50/pack

 
$669.74
   
Pack of 50
 
   
 

Hitachi M4 SEM mounts are made from lathe finished ultra-pure aluminum for scanning electron microscopy.

Surface: 1" (25 mm)
Height: 5/8" (16 mm)

 
  • Description:
    SEM Mount Hitachi M4 Aluminium 25 mm
  • Size:
    50/pack

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.