MeshWell Plates, Electron Microscopy Sciences
Product Details & Documents
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500 µm opening.
- Available with 6, 12, and 24 well configurations
- Also able to process tissue slices with Reagent Trays
Fits into the 6, 12 and 24 Costar/Corning Cell Culture Clusters for processing tissue slices.
Ordering information: The MeshWell Plate set includes one MeshWell and one Cluster plate.
Specifications
- Description:Plate 24–well with Tray and Lid
- Cat. No.:103306-230