Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Section Block, Electron Microscopy Sciences

Section Block, Electron Microscopy Sciences

Section Block, 125 mL
Catalog # 103306-198
Supplier #:  62711
Section Block, Electron Microscopy Sciences
Catalog # 103306-198
CAS Number:  
Section Block, Electron Microscopy Sciences

Section Block, 125 mL

CAS Number:  
Price
$492.21
Log in for availability
Each (125ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Blocking solutions based on chemically modified and fragmented ultra-pure casein. Effectively reduces unwanted binding of primary antibody and conjugates use to charged surface of the slide and tissue section.

Greatly reduces non-specific binding while preserving the specific reaction, by saturating potential non-specific protein-protein interactions.

Recommended for research and diagnostic pathology, especially for retrieved sections and polyclonal antibodies.

Specifications

Specifications

  • Description:
    Section Block
  • Size:
    125 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.