Orders LinkContactUsLinkComponent
Forensic Field Sampler, Electron Microscopy Sciences
Forensic Field Sampler, Electron Microscopy Sciences
Pin Stub with Adhesive Carbon Tab, 6 mm
Catalog # 103306-184
Supplier #:  76480-35
Forensic Field Sampler, Electron Microscopy Sciences
Catalog # 103306-184
Forensic Field Sampler, Electron Microscopy Sciences
Forensic Field Sampler, Electron Microscopy Sciences
Pin Stub with Adhesive Carbon Tab, 6 mm
Price
$410.13
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Collect forensic evidence with minimum interference and/or contamination from the sampler.
  • Gunshot residue (GSR)
  • Powder and fiber samples
  • Paint chips
  • Particle sampling
  • Glass fragments
Glass specimen vial (25 OD x 55 mm high) with a high-purity certified 12.7 mm SEM pin stub specimen mount, both with and without an adhesive carbon conductive tab, mounted into the plastic cap
Choice of standard 12.7 mm SEM pin mount with 8 mm pin or 12.7 mm SEM pin mount with shorter 6 mm pin (ZEISS/LEO). Ideal for SEM specimen preparation on the most frequently used SEM pin stubs, facilitating easy carbon coating for SEM/EDX investigation. The Forensic Field Sampler can be securely stored in the glass specimen vial and easily shipped with the cap holding the sampler stub in place. Adhesive carbon tabs secure the evidence material.
This Forensic Field Sampler has been designed to collect forensic evidence with minimum interference and/or contamination from the sampler. Three versions are available: without adhesive carbon tab, with ready-to-use adhesive carbon tab, or with covered carbon tab (plastic cover needs to be removed prior to use).
Specifications
  • Description:
    Pin Stub with Adhesive Carbon Tab
  • Pin size:
    6 mm