Orders LinkContactUsLinkComponent
Orbital Specimen Holder, Electron Microscopy Sciences
Orbital Specimen Holder, Electron Microscopy Sciences
Orbital Specimen Holder M4 Mount
Catalog # 103306-180
Supplier #:  75912
Orbital Specimen Holder, Electron Microscopy Sciences
Catalog # 103306-180
Orbital Specimen Holder, Electron Microscopy Sciences
Orbital Specimen Holder, Electron Microscopy Sciences
Orbital Specimen Holder M4 Mount
Price
$391.11
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Unique orbital specimen holder for a spin and raise SEM images to a whole new level.
  • Rotate 360° around the axis perpendicular to the electron beam
  • Position the sample with turn of a screw
  • Flexibile and superior control
The pot is coated to reduce noise, producing a black background behind the subject so every detail stands out, allowing the capture of stunning, clear images.
Specifications
  • Description:
    Orbital Specimen Holder M4 Mount