Orders LinkContactUsLinkComponent
Saws, Electron Microscopy Sciences
Saws, Electron Microscopy Sciences
Satterlee Saw, 292 mm Long
Catalog # 103306-032
Supplier #:  70566-31
Saws, Electron Microscopy Sciences
Catalog # 103306-032
Saws, Electron Microscopy Sciences
Saws, Electron Microscopy Sciences
Satterlee Saw, 292 mm Long
Price
$223.26
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Three styles of manual bone saws are available.
  • Broad bladed Satterlee Saw
  • Thinner rounded top Charriere Saw
  • Solid handle Langenbech Metacarpal Saw
The broad bladed Satterlee Saw (A), the thinner rounded tip Charriere Saw (B), and the solid handle Langenbech Metacarpal Saw (C). Each saw has serrated teeth.
Specifications
  • Description:
    Satterlee Saw, 292 mm Long